Product Details

SNP ID
rs139031066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78789733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTACAGTGAAGGCCAGCGGCAGGCC[C/T]GTGTCCTCGTTGATGCTGCAGCTGC
Phenotype
MIM: 605009
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAMTS7 PubMed Links

Gene Details

Gene
ADAMTS7
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014272.4 1377 Silent Mutation ACA,ACG T378T NP_055087.2
XM_005254137.3 1377 Silent Mutation ACA,ACG T378T XP_005254194.1
XM_011521166.2 1377 Intron XP_011519468.1

View Full Product Details