Product Details

SNP ID
rs139844938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:98437498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCCATCTTTGTAATGGGTGCAA[A/G]AGAGATGAAAACTCCAGTGGAACAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM169B PubMed Links

Gene Details

Gene
FAM169B
Gene Name
family with sequence similarity 169 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182562.2 2962 UTR 3 NP_872368.2

View Full Product Details