Product Details

SNP ID
rs143806224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75206106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGTGATGTATGGCAAGCTGCCC[C/T]GCTTAGAGACAGACTCCGGGCTCGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C15orf39 PubMed Links

Gene Details

Gene
C15orf39
Gene Name
chromosome 15 open reading frame 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015492.4 288 Missense Mutation CGC,TGC R20C NP_056307.2

View Full Product Details