Product Details

SNP ID
rs144292587
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64395431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGGCAGAAATCAGGCGACCATC[A/T]AAAGCGGGGTAGGAAGAAAGGGAGA
Phenotype
MIM: 610696 MIM: 604501
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KIAA0101 PubMed Links

Gene Details

Gene
KIAA0101
Gene Name
KIAA0101
There are no transcripts associated with this gene.

Gene
TRIP4
Gene Name
thyroid hormone receptor interactor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321924.1 365 UTR 5 NP_001308853.1
NM_016213.4 365 Missense Mutation CAA,CTA Q102L NP_057297.2

View Full Product Details