Product Details

SNP ID
rs145116014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:89785463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCAGGGCCCGGGTGCCTGAGCC[A/G]AAGCCTGTTTCCTCGTTGTCCTTCT
Phenotype
MIM: 151530 MIM: 605195
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANPEP PubMed Links

Gene Details

Gene
ANPEP
Gene Name
alanyl aminopeptidase, membrane
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001150.2 3082 Silent Mutation TTC,TTT F930F NP_001141.2
XM_005254892.4 3082 Silent Mutation TTC,TTT F930F XP_005254949.1
XM_011521473.1 3082 Silent Mutation TTC,TTT F930F XP_011519775.1
Gene
MESP2
Gene Name
mesoderm posterior bHLH transcription factor 2
There are no transcripts associated with this gene.

View Full Product Details