Product Details

SNP ID
rs146207678
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:38696628 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATATGGAGCTACAAGGGGCCCA[A/G]GAGGACCTGGGCATTTCCCTCTCTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C15orf53 PubMed Links

Gene Details

Gene
C15orf53
Gene Name
chromosome 15 open reading frame 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207444.2 31 Silent Mutation CAA,CAG Q7Q NP_997327.1

View Full Product Details