Product Details

SNP ID
rs147496975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64387894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGCTGGGGCGGTGTCCGGGGAG[C/T]CGCTGGTGCACTGGTGCACCCAGCA
Phenotype
MIM: 610696 MIM: 604501
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA0101 PubMed Links

Gene Details

Gene
KIAA0101
Gene Name
KIAA0101
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029989.2 91 Intron NP_001025160.1
NM_014736.5 91 Intron NP_055551.1
Gene
TRIP4
Gene Name
thyroid hormone receptor interactor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321924.1 91 UTR 5 NP_001308853.1
NM_016213.4 91 Missense Mutation CCG,TCG P11S NP_057297.2

View Full Product Details