Product Details

SNP ID
rs149590841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:63597724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCACGAGAGCCTGGTCAATGATT[G/T]CCTCCTCCGGGTGTGCGACAGGTAG
Phenotype
MIM: 609088 MIM: 604728
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FBXL22 PubMed Links

Gene Details

Gene
FBXL22
Gene Name
F-box and leucine rich repeat protein 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203373.2 334 Missense Mutation TGC,TTC C111F NP_976307.2
XM_005254320.3 334 Missense Mutation TGC,TTC C111F XP_005254377.1
XM_006720478.1 334 UTR 5 XP_006720541.1
XM_011521468.2 334 UTR 5 XP_011519770.1
XM_011521469.1 334 UTR 5 XP_011519771.1
XM_011521472.2 334 UTR 5 XP_011519774.1
XM_017022086.1 334 Missense Mutation TGC,TTC C111F XP_016877575.1
Gene
USP3
Gene Name
ubiquitin specific peptidase 3
There are no transcripts associated with this gene.

Gene
USP3-AS1
Gene Name
USP3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details