Product Details

SNP ID
rs151188660
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74719949 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGGGCAGAGGAATGTGATGTTC[A/C]CAGGAACTGTGTCCTAGACCCATAG
Phenotype
MIM: 108330
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CYP1A1 PubMed Links

Gene Details

Gene
CYP1A1
Gene Name
cytochrome P450 family 1 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000499.4 2278 UTR 3 NP_000490.1
NM_001319216.1 2278 UTR 3 NP_001306145.1
NM_001319217.1 2278 UTR 3 NP_001306146.1
XM_017021953.1 2278 UTR 3 XP_016877442.1

View Full Product Details