Product Details

SNP ID
rs138915039
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4736617 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCAAGGCTGTCAAAGACCAGCTC[C/T]CGTCTCTGGACTCAGACTCCCCTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKS3 PubMed Links

Gene Details

Gene
ANKS3
Gene Name
ankyrin repeat and sterile alpha motif domain containing 3
There are no transcripts associated with this gene.

Gene
C16orf71
Gene Name
chromosome 16 open reading frame 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139170.2 183 Missense Mutation CCG,TCG P35S NP_631909.2
XM_005255144.3 183 Intron XP_005255201.1
XM_017022975.1 183 Missense Mutation CCG,TCG P58S XP_016878464.1
XM_017022976.1 183 Missense Mutation CCG,TCG P35S XP_016878465.1
Gene
ZNF500
Gene Name
zinc finger protein 500
There are no transcripts associated with this gene.

View Full Product Details