Product Details

SNP ID
rs140663234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1511027 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCTGTTGTGGCGGACCTGCTCGG[C/G]GACGGTGCGTGGCAGTGGGAGACCC
Phenotype
MIM: 614620 MIM: 611140
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IFT140 PubMed Links

Gene Details

Gene
IFT140
Gene Name
intraflagellar transport 140
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014714.3 3388 Missense Mutation CCC,GCC P1436A NP_055529.2
XM_005255725.4 3388 Intron XP_005255782.1
XM_005255726.3 3388 Intron XP_005255783.1
XM_006720990.3 3388 Missense Mutation CCC,GCC P1436A XP_006721053.1
XM_006720991.3 3388 Missense Mutation CCC,GCC P1436A XP_006721054.1
XM_006720992.3 3388 Missense Mutation CCC,GCC P647A XP_006721055.1
XM_011522766.2 3388 Missense Mutation CCC,GCC P1354A XP_011521068.1
XM_011522767.2 3388 Missense Mutation CCC,GCC P1111A XP_011521069.1
XM_011522769.2 3388 Intron XP_011521071.1
XM_011522771.2 3388 Intron XP_011521073.1
XM_011522772.2 3388 Intron XP_011521074.1
XM_017023910.1 3388 Missense Mutation CCC,GCC P1436A XP_016879399.1
XM_017023911.1 3388 Missense Mutation CCC,GCC P831A XP_016879400.1
Gene
TELO2
Gene Name
telomere maintenance 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016111.3 3388 Intron NP_057195.2
XM_011522773.2 3388 Intron XP_011521075.1
XM_011522774.2 3388 Intron XP_011521076.1
XM_011522775.2 3388 Intron XP_011521077.1
XM_011522776.2 3388 Intron XP_011521078.1
XM_011522777.2 3388 Intron XP_011521079.1
XM_011522778.2 3388 Intron XP_011521080.1
XM_017023914.1 3388 Intron XP_016879403.1

View Full Product Details