Product Details

SNP ID
rs141446517
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29663850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGTCTCTACTAGCGAGCCCCTGA[A/G]CTCAAAGATGTACACCACTTCAATA
Phenotype
MIM: 606248 MIM: 182160
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
QPRT PubMed Links

Gene Details

Gene
QPRT
Gene Name
quinolinate phosphoribosyltransferase
There are no transcripts associated with this gene.

Gene
SPN
Gene Name
sialophorin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030288.2 287 Missense Mutation AAC,AGC N41S NP_001025459.1
NM_003123.4 287 Missense Mutation AAC,AGC N41S NP_003114.1

View Full Product Details