Product Details

SNP ID
rs143906081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23587233 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATCTTGTCATAGAGTTTCAATAC[A/G]TAAAGAACACGGTCCTGGATGCCCT
Phenotype
MIM: 603836
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NDUFAB1 PubMed Links

Gene Details

Gene
NDUFAB1
Gene Name
NADH:ubiquinone oxidoreductase subunit AB1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005003.2 387 Silent Mutation TAC,TAT Y85Y NP_004994.1
XM_011545856.2 387 Silent Mutation TAC,TAT Y116Y XP_011544158.2

View Full Product Details