Product Details

SNP ID
rs144051380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23581966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCCAGCAGGCTGGGCTTACCTTG[A/G]CTGAGTAATTGAGGGAGACAGAGGT
Phenotype
MIM: 603836
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NDUFAB1 PubMed Links

Gene Details

Gene
NDUFAB1
Gene Name
NADH:ubiquinone oxidoreductase subunit AB1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005003.2 Intron NP_004994.1
XM_011545856.2 Intron XP_011544158.2
Gene
UBFD1
Gene Name
ubiquitin family domain containing 1
There are no transcripts associated with this gene.

View Full Product Details