Product Details

SNP ID
rs144997996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:46662398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTCTCTTCACTGAAGCACTTCA[A/T]CCTTTCAAAAGTGCCAATGATCAAG
Phenotype
MIM: 601501
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
VPS35 PubMed Links

Gene Details

Gene
VPS35
Gene Name
VPS35 retromer complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018206.5 2011 Missense Mutation ATG,TTG M638L NP_060676.2
XM_005256045.3 2011 Missense Mutation ATG,TTG M571L XP_005256102.1
XM_011523227.2 2011 Missense Mutation ATG,TTG M609L XP_011521529.1

View Full Product Details