Product Details

SNP ID
rs145033509
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:46661784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATTTCTATAAAAAGCTGCACTTG[C/T]AGAGAGGGGTCCATGCACTGATTTG
Phenotype
MIM: 601501
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VPS35 PubMed Links

Gene Details

Gene
VPS35
Gene Name
VPS35 retromer complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018206.5 2244 Silent Mutation CTA,CTG L715L NP_060676.2
XM_005256045.3 2244 Silent Mutation CTA,CTG L648L XP_005256102.1
XM_011523227.2 2244 Silent Mutation CTA,CTG L686L XP_011521529.1

View Full Product Details