Product Details

SNP ID
rs145286621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:11752763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACCTTCTCTTTGTGCTTCTCGG[A/C]GGAGATGTGGCCCTGCCACTGCTTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TXNDC11 PubMed Links

Gene Details

Gene
TXNDC11
Gene Name
thioredoxin domain containing 11
There are no transcripts associated with this gene.

Gene
ZC3H7A
Gene Name
zinc finger CCCH-type containing 7A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014153.3 2594 Missense Mutation GCC,TCC A878S NP_054872.2
XM_006720877.2 2594 Missense Mutation GCC,TCC A878S XP_006720940.1
XM_011522463.1 2594 Missense Mutation GCC,TCC A878S XP_011520765.1
XM_011522464.2 2594 Missense Mutation GCC,TCC A639S XP_011520766.1
XM_017023174.1 2594 Missense Mutation GCC,TCC A777S XP_016878663.1
XM_017023175.1 2594 Missense Mutation GCC,TCC A487S XP_016878664.1

View Full Product Details