Product Details

SNP ID
rs146091204
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28880946 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGTTTGAGGAAGGTGAAGAGA[C/T]CATCACTGCCTTTGTTGAACCCTTT
Phenotype
MIM: 108730 MIM: 608937
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP2A1 PubMed Links

Gene Details

Gene
ATP2A1
Gene Name
ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286075.1 435 UTR 5 NP_001273004.1
NM_004320.4 435 Missense Mutation ACC,ATC T84I NP_004311.1
NM_173201.3 435 Missense Mutation ACC,ATC T84I NP_775293.1
Gene
ATP2A1-AS1
Gene Name
ATP2A1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
SH2B1
Gene Name
SH2B adaptor protein 1
There are no transcripts associated with this gene.

View Full Product Details