Product Details

SNP ID
rs146552672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:11679353 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGGCTTTCAGCTCCATCCCTGC[C/T]CTCCAGTTTCCTCTCCATGGTCGCC
Phenotype
MIM: 603032
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNN PubMed Links

Gene Details

Gene
SNN
Gene Name
stannin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003498.5 2704 Intron NP_003489.1
XM_017023741.1 2704 Intron XP_016879230.1
XM_017023742.1 2704 Intron XP_016879231.1
Gene
TXNDC11
Gene Name
thioredoxin domain containing 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303447.1 2704 Missense Mutation AGC,GGC S934G NP_001290376.1
NM_001324022.1 2704 Missense Mutation AGC,GGC S693G NP_001310951.1
NM_001324024.1 2704 Missense Mutation AGC,GGC S455G NP_001310953.1
NM_001324025.1 2704 Missense Mutation AGC,GGC S455G NP_001310954.1
NM_015914.6 2704 Missense Mutation AGC,GGC S907G NP_056998.4
XM_011522515.2 2704 Missense Mutation AGC,GGC S947G XP_011520817.1
XM_011522516.2 2704 Missense Mutation AGC,GGC S733G XP_011520818.1
XM_017023268.1 2704 Missense Mutation AGC,GGC S946G XP_016878757.1

View Full Product Details