Product Details
- SNP ID
-
rs149102650
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:23755881 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTCCAGCAGATAGGGGCGCTCGCC[A/G]TGAACCCCCTGGGAGACCGAATTAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CHP2
PubMed Links
Gene Details
- Gene
- CHP2
- Gene Name
- calcineurin like EF-hand protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022097.3 |
598 |
Missense Mutation |
ATG,GTG |
M59V |
NP_071380.1 |
View Full Product Details