Product Details
- SNP ID
-
rs149698079
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:74716465 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATGGGTCATGTCATAGAGGACGTA[A/G]CCCAGGAGGCCCCCCGCAAACACAG
- Phenotype
-
MIM: 611026
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FA2H
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11554620] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FA2H
- Gene Name
- fatty acid 2-hydroxylase
View Full Product Details