Product Details

SNP ID
rs137974930
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75890928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGTGTACAGGGGCTGGGTCTGGG[G/T]CTCCAGGCTGTAGAAGGTGAGGCAG
Phenotype
MIM: 611844
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MRPL38 PubMed Links

Gene Details

Gene
MRPL38
Gene Name
mitochondrial ribosomal protein L38
There are no transcripts associated with this gene.

Gene
TRIM65
Gene Name
tripartite motif containing 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256124.1 1442 Missense Mutation ACC,CCC T447P NP_001243053.1
NM_173547.3 1442 Missense Mutation ACC,CCC T469P NP_775818.2
XM_006721760.3 1442 Missense Mutation ACC,CCC T482P XP_006721823.1
XM_006721761.3 1442 Missense Mutation ACC,CCC T460P XP_006721824.1
XM_011524499.2 1442 Intron XP_011522801.1
XM_011524500.2 1442 Intron XP_011522802.1
XM_011524501.2 1442 Intron XP_011522803.1
XM_011524502.2 1442 Intron XP_011522804.1
XM_011524503.2 1442 Intron XP_011522805.1
XM_017024343.1 1442 Intron XP_016879832.1
XM_017024344.1 1442 UTR 3 XP_016879833.1

View Full Product Details