Product Details

SNP ID
rs138597553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:67044705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGGTCCGCGTGACCCTCTTCTG[C/T]ATCCTGGCAGGCATCGTGCTGGCCA
Phenotype
MIM: 114209
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CACNG1 PubMed Links

Gene Details

Gene
CACNG1
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000727.3 170 Silent Mutation TGC,TGT C15C NP_000718.1

View Full Product Details