Product Details

SNP ID
rs139277891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78675448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCCCTCTGCGTCCTCGACCTCA[C/T]CTGTCCATCTCGCCCAGACAAGCAC
Phenotype
MIM: 182115
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYTH1 PubMed Links

Gene Details

Gene
CYTH1
Gene Name
cytohesin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292018.1 2356 UTR 3 NP_001278947.1
NM_001292019.1 2356 UTR 3 NP_001278948.1
NM_004762.3 2356 UTR 3 NP_004753.1
NM_017456.3 2356 UTR 3 NP_059430.2
XM_011525475.2 2356 UTR 3 XP_011523777.1
XM_011525476.2 2356 UTR 3 XP_011523778.1
XM_011525477.1 2356 UTR 3 XP_011523779.1
XM_011525478.1 2356 UTR 3 XP_011523780.1
XM_017025336.1 2356 UTR 3 XP_016880825.1
XM_017025337.1 2356 Intron XP_016880826.1
XM_017025338.1 2356 UTR 3 XP_016880827.1

View Full Product Details