Product Details

SNP ID
rs141452748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35435988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCCTGACTTGAACCACCTTGGTC[A/G]TGGTGTATTATCACTTTTACAAATT
Phenotype
MIM: 614955 MIM: 614957
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLFN12 PubMed Links

Gene Details

Gene
SLFN12
Gene Name
schlafen family member 12
There are no transcripts associated with this gene.

Gene
SLFN13
Gene Name
schlafen family member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144682.5 7577 UTR 3 NP_653283.3
XM_005257922.2 7577 Intron XP_005257979.1
XM_011524383.2 7577 Intron XP_011522685.1
XM_017024232.1 7577 Intron XP_016879721.1
XM_017024233.1 7577 Intron XP_016879722.1

View Full Product Details