Product Details

SNP ID
rs145164915
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40794000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTGGATCTGCGCCAGCTGCGTA[C/T]AGTAGTTGCTCTCGGTCTCTGTCAA
Phenotype
MIM: 616677
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KRT28 PubMed Links
Additional Information
For this assay, SNP(s) [rs2270154] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
KRT28
Gene Name
keratin 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181535.3 1091 Missense Mutation TAT,TGT Y342C NP_853513.2

View Full Product Details