Product Details
- SNP ID
-
rs145403707
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:82444498 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGTGAGGGGGTGGGAGAAGCCCGTC[A/G]CAAGCCGGAGCACCACCATGTAGCC
- Phenotype
-
MIM: 616864
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C17orf62
PubMed Links
Gene Details
- Gene
- C17orf62
- Gene Name
- chromosome 17 open reading frame 62
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001033046.3 |
557 |
Missense Mutation |
GCG,GTG |
A131V |
NP_001028218.1 |
NM_001100407.2 |
557 |
Missense Mutation |
GCG,GTG |
A131V |
NP_001093877.1 |
NM_001100408.2 |
557 |
Missense Mutation |
GCG,GTG |
A117V |
NP_001093878.1 |
NM_001193653.1 |
557 |
Missense Mutation |
GCG,GTG |
A131V |
NP_001180582.1 |
NM_001193654.1 |
557 |
Missense Mutation |
GCG,GTG |
A131V |
NP_001180583.1 |
NM_001193655.1 |
557 |
Missense Mutation |
GCG,GTG |
A131V |
NP_001180584.1 |
NM_001193657.1 |
557 |
Missense Mutation |
GCG,GTG |
A131V |
NP_001180586.1 |
XM_006722293.2 |
557 |
Missense Mutation |
GCG,GTG |
A117V |
XP_006722356.1 |
XM_011523606.2 |
557 |
Missense Mutation |
GCG,GTG |
A164V |
XP_011521908.1 |
XM_017025072.1 |
557 |
Missense Mutation |
GCG,GTG |
A164V |
XP_016880561.1 |
XM_017025073.1 |
557 |
Missense Mutation |
GCG,GTG |
A150V |
XP_016880562.1 |
XM_017025074.1 |
557 |
Missense Mutation |
GCG,GTG |
A131V |
XP_016880563.1 |
XM_017025075.1 |
557 |
Missense Mutation |
GCG,GTG |
A131V |
XP_016880564.1 |
XM_017025076.1 |
557 |
Missense Mutation |
GCG,GTG |
A131V |
XP_016880565.1 |
XM_017025077.1 |
557 |
Missense Mutation |
GCG,GTG |
A131V |
XP_016880566.1 |
XM_017025078.1 |
557 |
Missense Mutation |
GCG,GTG |
A117V |
XP_016880567.1 |
- Gene
- HEXDC
- Gene Name
- hexosaminidase D
There are no transcripts associated with this gene.
View Full Product Details