Product Details

SNP ID
rs145477224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75890818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTGGGGCCCCAGAGGGAACAC[A/G]GCCCCTGGCTGATGGCACAGGGTCA
Phenotype
MIM: 611844
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL38 PubMed Links

Gene Details

Gene
MRPL38
Gene Name
mitochondrial ribosomal protein L38
There are no transcripts associated with this gene.

Gene
TRIM65
Gene Name
tripartite motif containing 65
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256124.1 1552 Silent Mutation GCC,GCT A483A NP_001243053.1
NM_173547.3 1552 Silent Mutation GCC,GCT A505A NP_775818.2
XM_006721760.3 1552 Silent Mutation GCC,GCT A518A XP_006721823.1
XM_006721761.3 1552 Silent Mutation GCC,GCT A496A XP_006721824.1
XM_011524499.2 1552 Intron XP_011522801.1
XM_011524500.2 1552 Intron XP_011522802.1
XM_011524501.2 1552 Intron XP_011522803.1
XM_011524502.2 1552 Intron XP_011522804.1
XM_011524503.2 1552 Intron XP_011522805.1
XM_017024343.1 1552 Intron XP_016879832.1
XM_017024344.1 1552 UTR 3 XP_016879833.1

View Full Product Details