Product Details

SNP ID
rs147786222
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:754347 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGTAAATGGTATCCAGAAAGGCA[G/T]GTCTACGGTGGCATATGGGATTTCC
Phenotype
MIM: 606969
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DBIL5P PubMed Links

Gene Details

Gene
DBIL5P
Gene Name
diazepam binding inhibitor-like 5, pseudogene
There are no transcripts associated with this gene.

Gene
GEMIN4
Gene Name
gem nuclear organelle associated protein 4
There are no transcripts associated with this gene.

Gene
GLOD4
Gene Name
glyoxalase domain containing 4
There are no transcripts associated with this gene.

View Full Product Details