Product Details

SNP ID
rs148177663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:60222490 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTGGGACCATTTCATAGTCTCT[C/T]CCATGAATCAGCTGTGGAGTTCGTT
Phenotype
MIM: 607740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCARNA20 PubMed Links

Gene Details

Gene
SCARNA20
Gene Name
small Cajal body-specific RNA 20
There are no transcripts associated with this gene.

Gene
USP32
Gene Name
ubiquitin specific peptidase 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032582.3 1168 Silent Mutation GGA,GGG G556G NP_115971.2
XM_011525371.1 1168 Silent Mutation GGA,GGG G572G XP_011523673.1
XM_011525372.1 1168 Silent Mutation GGA,GGG G572G XP_011523674.1
XM_011525373.1 1168 Silent Mutation GGA,GGG G556G XP_011523675.1
XM_011525374.1 1168 Silent Mutation GGA,GGG G553G XP_011523676.1
XM_011525375.1 1168 Silent Mutation GGA,GGG G546G XP_011523677.1
XM_011525376.1 1168 Silent Mutation GGA,GGG G553G XP_011523678.1
XM_011525378.1 1168 Silent Mutation GGA,GGG G510G XP_011523680.1
XM_011525379.2 1168 Silent Mutation GGA,GGG G265G XP_011523681.1
XM_017025233.1 1168 Silent Mutation GGA,GGG G510G XP_016880722.1
XM_017025234.1 1168 Silent Mutation GGA,GGG G226G XP_016880723.1

View Full Product Details