Product Details

SNP ID
rs149157215
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42782916 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGGTCACCGAACTCATGACCTC[A/G]GGCACGCTCAAGACGTGAGCTCTGC
Phenotype
MIM: 610907 MIM: 601844
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VPS25 PubMed Links

Gene Details

Gene
VPS25
Gene Name
vacuolar protein sorting 25 homolog
There are no transcripts associated with this gene.

Gene
WNK4
Gene Name
WNK lysine deficient protein kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321299.1 845 UTR 5 NP_001308228.1
NM_032387.4 845 Silent Mutation TCA,TCG S259S NP_115763.2
XM_011525135.2 845 Silent Mutation TCA,TCG S259S XP_011523437.1
XM_017024962.1 845 Silent Mutation TCA,TCG S259S XP_016880451.1
XM_017024963.1 845 Silent Mutation TCA,TCG S259S XP_016880452.1
XM_017024964.1 845 Silent Mutation TCA,TCG S259S XP_016880453.1
XM_017024965.1 845 Silent Mutation TCA,TCG S259S XP_016880454.1
XM_017024966.1 845 Intron XP_016880455.1

View Full Product Details