Product Details

SNP ID
rs149790109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8757385 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTGTCCTCGGGCTCAGGGGCGC[A/G]CTCCAACTCCAGCTCCTCCTCCAGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPDYE4 PubMed Links

Gene Details

Gene
SPDYE4
Gene Name
speedy/RINGO cell cycle regulator family member E4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128076.1 1263 Missense Mutation CGC,TGC R73C NP_001121548.1
XM_005256632.4 1263 Missense Mutation CGC,TGC R73C XP_005256689.1
XM_005256633.4 1263 Missense Mutation CGC,TGC R73C XP_005256690.1

View Full Product Details