Product Details

SNP ID
rs138447200
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:67511144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTAGAGGTGGCAAACAATATTTG[G/T]TTTAAACTAGCAGGAGACAAAGGAA
Phenotype
MIM: 611125
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DSEL PubMed Links

Gene Details

Gene
DSEL
Gene Name
dermatan sulfate epimerase-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032160.2 4719 Silent Mutation NP_115536.1
Gene
LOC643542
Gene Name
uncharacterized LOC643542
There are no transcripts associated with this gene.

View Full Product Details