Product Details
- SNP ID
-
rs141737883
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:46477832 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTTGTCACGATGACTTCGTACTT[C/G]ACGGGCACCAGGCTCTGGACCTTGC
- Phenotype
-
MIM: 613072
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LOXHD1
PubMed Links
Gene Details
- Gene
- LOXHD1
- Gene Name
- lipoxygenase homology domains 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001145472.2 |
3082 |
Silent Mutation |
GTC,GTG |
V1043V |
NP_001138944.1 |
NM_001145473.2 |
3082 |
Silent Mutation |
GTC,GTG |
V393V |
NP_001138945.1 |
NM_001173129.1 |
3082 |
Silent Mutation |
GTC,GTG |
V393V |
NP_001166600.1 |
NM_001308013.1 |
3082 |
Silent Mutation |
GTC,GTG |
V947V |
NP_001294942.1 |
NM_144612.6 |
3082 |
Silent Mutation |
GTC,GTG |
V2092V |
NP_653213.6 |
XM_006722388.3 |
3082 |
Silent Mutation |
GTC,GTG |
V1087V |
XP_006722451.1 |
XM_006722389.3 |
3082 |
Silent Mutation |
GTC,GTG |
V1043V |
XP_006722452.1 |
XM_006722390.3 |
3082 |
Silent Mutation |
GTC,GTG |
V1043V |
XP_006722453.1 |
XM_006722391.3 |
3082 |
Silent Mutation |
GTC,GTG |
V1025V |
XP_006722454.1 |
XM_011525804.2 |
3082 |
Silent Mutation |
GTC,GTG |
V1541V |
XP_011524106.1 |
XM_011525807.2 |
3082 |
Silent Mutation |
GTC,GTG |
V947V |
XP_011524109.1 |
XM_011525810.2 |
3082 |
Silent Mutation |
GTC,GTG |
V410V |
XP_011524112.1 |
XM_011525811.2 |
3082 |
Silent Mutation |
GTC,GTG |
V393V |
XP_011524113.1 |
XM_017025540.1 |
3082 |
Silent Mutation |
GTC,GTG |
V1806V |
XP_016881029.1 |
XM_017025541.1 |
3082 |
Silent Mutation |
GTC,GTG |
V1835V |
XP_016881030.1 |
XM_017025542.1 |
3082 |
Intron |
|
|
XP_016881031.1 |
XM_017025543.1 |
3082 |
Intron |
|
|
XP_016881032.1 |
XM_017025544.1 |
3082 |
Intron |
|
|
XP_016881033.1 |
XM_017025545.1 |
3082 |
Intron |
|
|
XP_016881034.1 |
XM_017025546.1 |
3082 |
Intron |
|
|
XP_016881035.1 |
XM_017025547.1 |
3082 |
Intron |
|
|
XP_016881036.1 |
XM_017025548.1 |
3082 |
Silent Mutation |
GTC,GTG |
V1948V |
XP_016881037.1 |
View Full Product Details