Product Details
- SNP ID
-
rs142875983
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:60371594 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGGGGCCCAGCAGACAACAAAGAC[A/G]CCAATCAGGATGGTCAAGGTAATCG
- Phenotype
-
MIM: 155541
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MC4R
PubMed Links
Gene Details
- Gene
- MC4R
- Gene Name
- melanocortin 4 receptor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005912.2 |
1175 |
Silent Mutation |
GGC,GGT |
G252G |
NP_005903.2 |
View Full Product Details