Product Details

SNP ID
rs147575731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:9105826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTATTGTGTATTTTTTCATTCAT[A/C]GAGGATATTTTCCTTTATGTCTTTA
Phenotype
MIM: 600532
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NDUFV2 PubMed Links

Gene Details

Gene
NDUFV2
Gene Name
NADH:ubiquinone oxidoreductase core subunit V2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021074.4 Intron NP_066552.2
XM_017025782.1 Intron XP_016881271.1

View Full Product Details