Product Details

SNP ID
rs148630515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:74148960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGGCTTCAATATCAAAAGAAAA[C/T]GCTGCGACCTAGATGTATTCTTGGA
Phenotype
MIM: 609093 MIM: 615180
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXO15 PubMed Links

Gene Details

Gene
FBXO15
Gene Name
F-box protein 15
There are no transcripts associated with this gene.

Gene
TIMM21
Gene Name
translocase of inner mitochondrial membrane 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014177.2 434 Missense Mutation ACG,ATG T51M NP_054896.2
XM_005266689.3 434 UTR 5 XP_005266746.1

View Full Product Details