Product Details

SNP ID
rs138040696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:36112882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGCACCTCAAGTGCCACAGCCC[C/T]GTGCGCCGCCACCTGTGCCGCTGTT
Phenotype
MIM: 616442 MIM: 180662 MIM: 601303 MIM: 613583
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OVOL3 PubMed Links

Gene Details

Gene
OVOL3
Gene Name
ovo like zinc finger 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302757.1 282 Silent Mutation CCC,CCT P94P NP_001289686.1
XM_011527251.2 282 Silent Mutation CCC,CCT P11P XP_011525553.1
XM_017027190.1 282 Silent Mutation CCC,CCT P117P XP_016882679.1
XM_017027191.1 282 Silent Mutation CCC,CCT P93P XP_016882680.1
Gene
POLR2I
Gene Name
RNA polymerase II subunit I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006233.4 282 Intron NP_006224.1
Gene
TBCB
Gene Name
tubulin folding cofactor B
There are no transcripts associated with this gene.

Gene
WDR62
Gene Name
WD repeat domain 62
There are no transcripts associated with this gene.

View Full Product Details