Product Details

SNP ID
rs140006700
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9102318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAATGGCACACCACAGAAGAGAA[A/G]GTATCCTCCCTATGAACTTCCTCAA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
OR1M1 PubMed Links

Gene Details

Gene
OR1M1
Gene Name
olfactory receptor family 1 subfamily M member 1
There are no transcripts associated with this gene.

Gene
OR7G2
Gene Name
olfactory receptor family 7 subfamily G member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005193.1 989 Missense Mutation CCT,CTT P330L NP_001005193.1

View Full Product Details