Product Details

SNP ID
rs141158542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45163992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAGAGGGCGCCGCGCAGGAGGCC[A/G]CAGGTGAAGGCCAGGAACTGAGGAG
Phenotype
MIM: 610396
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NKPD1 PubMed Links

Gene Details

Gene
NKPD1
Gene Name
NTPase, KAP family P-loop domain containing 1
There are no transcripts associated with this gene.

Gene
TRAPPC6A
Gene Name
trafficking protein particle complex 6A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270891.1 397 Silent Mutation TGC,TGT C124C NP_001257820.1
NM_001270892.1 397 Missense Mutation CGG,TGG R116W NP_001257821.1
NM_001270893.1 397 Missense Mutation CGG,TGG R102W NP_001257822.1
NM_024108.2 397 Silent Mutation TGC,TGT C138C NP_077013.1
XM_017027295.1 397 Missense Mutation CGG,TGG R151W XP_016882784.1

View Full Product Details