Product Details

SNP ID
rs141686756
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7619775 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCTCCAGGTCCATCTCGTCCTC[A/G]TCCTCGTCCTCGCCCAGCTGGATCT
Phenotype
MIM: 612685 MIM: 610850
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CAMSAP3 PubMed Links

Gene Details

Gene
CAMSAP3
Gene Name
calmodulin regulated spectrin associated protein family member 3
There are no transcripts associated with this gene.

Gene
MIR6792
Gene Name
microRNA 6792
There are no transcripts associated with this gene.

Gene
XAB2
Gene Name
XPA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020196.2 2504 Silent Mutation GAC,GAT D826D NP_064581.2

View Full Product Details