Product Details

SNP ID
rs143089101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53259269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACAAACAGCAGGTACAACACAT[A/C]TGTAGGAACAATCTCTACCCCAACT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
VN1R2 PubMed Links

Gene Details

Gene
VN1R2
Gene Name
vomeronasal 1 receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173856.2 978 Silent Mutation ATA,ATC I298I NP_776255.2
Gene
VN1R4
Gene Name
vomeronasal 1 receptor 4
There are no transcripts associated with this gene.

Gene
ZNF677
Gene Name
zinc finger protein 677
There are no transcripts associated with this gene.

View Full Product Details