Product Details

SNP ID
rs143291075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37153503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGAATATCTTTTCAAATTTGGC[A/G]CATTCATAAGCTTTCTCCCCAGGAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF585A PubMed Links

Gene Details

Gene
ZNF585A
Gene Name
zinc finger protein 585A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288800.1 655 Silent Mutation TGC,TGT C132C NP_001275729.1
NM_152655.3 655 Silent Mutation TGC,TGT C77C NP_689868.1
NM_199126.2 655 Silent Mutation TGC,TGT C77C NP_954577.1

View Full Product Details