Product Details

SNP ID
rs144099316
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57590021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCTCTGCTTTCAGGTTGTGGCCA[A/T]GGAACAGAGGATGAAGAGACACCTT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZIK1 PubMed Links

Gene Details

Gene
ZIK1
Gene Name
zinc finger protein interacting with K protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010879.3 552 Silent Mutation CAA,CAT Q70H NP_001010879.2
NM_001321145.1 552 Silent Mutation CAA,CAT Q57H NP_001308074.1
NM_001321146.1 552 Silent Mutation CAA,CAT Q15H NP_001308075.1
NM_001321147.1 552 UTR 5 NP_001308076.1
XM_011526761.1 552 Missense Mutation CAA,CAT Q56H XP_011525063.1
XM_011526762.1 552 Missense Mutation CAA,CAT Q51H XP_011525064.1
XM_011526763.1 552 Missense Mutation CAA,CAT Q43H XP_011525065.1
XM_011526764.1 552 Missense Mutation CAA,CAT Q37H XP_011525066.1
XM_011526765.2 552 Missense Mutation CAA,CAT Q34H XP_011525067.1
XM_011526766.2 552 Missense Mutation CAA,CAT Q15H XP_011525068.1
XM_011526767.2 552 UTR 5 XP_011525069.1
Gene
ZNF530
Gene Name
zinc finger protein 530
There are no transcripts associated with this gene.

View Full Product Details