Product Details
- SNP ID
-
rs145189460
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:43913437 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTGATGAAGGAAAGTCCTTGTCAC[C/G]CTCATCATCAGCATGGACTCGCTGA
- Phenotype
-
MIM: 194554
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF45
PubMed Links
Gene Details
- Gene
- ZNF45
- Gene Name
- zinc finger protein 45
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003425.3 |
3153 |
Missense Mutation |
CGT,GGT |
R667G |
NP_003416.1 |
XM_011527267.1 |
3153 |
Missense Mutation |
CGT,GGT |
R667G |
XP_011525569.1 |
XM_011527269.1 |
3153 |
Missense Mutation |
CGT,GGT |
R667G |
XP_011525571.1 |
XM_011527271.1 |
3153 |
Missense Mutation |
CGT,GGT |
R667G |
XP_011525573.1 |
XM_011527273.1 |
3153 |
Missense Mutation |
CGT,GGT |
R667G |
XP_011525575.1 |
XM_017027217.1 |
3153 |
Missense Mutation |
CGT,GGT |
R667G |
XP_016882706.1 |
XM_017027218.1 |
3153 |
Missense Mutation |
CGT,GGT |
R667G |
XP_016882707.1 |
XM_017027219.1 |
3153 |
Missense Mutation |
CGT,GGT |
R667G |
XP_016882708.1 |
XM_017027220.1 |
3153 |
Missense Mutation |
CGT,GGT |
R667G |
XP_016882709.1 |
XM_017027221.1 |
3153 |
Missense Mutation |
CGT,GGT |
R667G |
XP_016882710.1 |
XM_017027222.1 |
3153 |
Missense Mutation |
CGT,GGT |
R667G |
XP_016882711.1 |
XM_017027223.1 |
3153 |
Missense Mutation |
CGT,GGT |
R667G |
XP_016882712.1 |
XM_017027224.1 |
3153 |
Missense Mutation |
CGT,GGT |
R667G |
XP_016882713.1 |
XM_017027225.1 |
3153 |
Missense Mutation |
CGT,GGT |
R667G |
XP_016882714.1 |
XM_017027226.1 |
3153 |
Missense Mutation |
CGT,GGT |
R667G |
XP_016882715.1 |
XM_017027227.1 |
3153 |
Missense Mutation |
CGT,GGT |
R667G |
XP_016882716.1 |
XM_017027228.1 |
3153 |
Missense Mutation |
CGT,GGT |
R631G |
XP_016882717.1 |
XM_017027229.1 |
3153 |
Missense Mutation |
CGT,GGT |
R631G |
XP_016882718.1 |
XM_017027230.1 |
3153 |
Missense Mutation |
CGT,GGT |
R631G |
XP_016882719.1 |
View Full Product Details