Product Details

SNP ID
rs145588887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15728591 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCATTTTTTATGTCATGTGCCAC[C/G]TCTGTTGAAGTTGGCCTGTGGAAAT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR10H2 PubMed Links

Gene Details

Gene
OR10H2
Gene Name
olfactory receptor family 10 subfamily H member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013939.2 568 Missense Mutation CCT,CGT P183R NP_039227.1

View Full Product Details