Product Details

SNP ID
rs145619119
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15728671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGTATGTATCATGGCACTGCTG[A/G]GCTGTTTTCTCCTCATCCTCCTCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR10H2 PubMed Links

Gene Details

Gene
OR10H2
Gene Name
olfactory receptor family 10 subfamily H member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013939.2 648 Missense Mutation AGC,GGC S210G NP_039227.1

View Full Product Details