Product Details

SNP ID
rs146123396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41621724 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACCCCAACCAGGACCCCAGTCA[C/T]GATGCCAGCGACGGCCCCCACAGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEACAM4 PubMed Links

Gene Details

Gene
CEACAM4
Gene Name
carcinoembryonic antigen related cell adhesion molecule 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001817.3 580 Intron NP_001808.2
XM_006722977.3 580 Intron XP_006723040.1
XM_011526343.1 580 Intron XP_011524645.1
XM_011526345.2 580 Intron XP_011524647.1
XM_011526348.2 580 Missense Mutation ATG,GTG M157V XP_011524650.1
XM_011526349.2 580 Intron XP_011524651.1
XM_011526350.2 580 Missense Mutation ATG,GTG M157V XP_011524652.1
XM_011526352.1 580 Missense Mutation ATG,GTG M157V XP_011524654.1
XM_011526354.1 580 Intron XP_011524656.1
XM_011526357.2 580 Intron XP_011524659.1
XM_017026199.1 580 Intron XP_016881688.1
XM_017026200.1 580 Missense Mutation ATG,GTG M157V XP_016881689.1
XM_017026201.1 580 Intron XP_016881690.1
XM_017026202.1 580 Intron XP_016881691.1
XM_017026203.1 580 Intron XP_016881692.1
XM_017026204.1 580 Intron XP_016881693.1
XM_017026205.1 580 Missense Mutation ATG,GTG M157V XP_016881694.1
XM_017026206.1 580 Missense Mutation ATG,GTG M157V XP_016881695.1
XM_017026207.1 580 Intron XP_016881696.1
XM_017026208.1 580 Intron XP_016881697.1
XM_017026209.1 580 Missense Mutation ATG,GTG M157V XP_016881698.1
XM_017026210.1 580 Missense Mutation ATG,GTG M157V XP_016881699.1
XM_017026211.1 580 Intron XP_016881700.1
XM_017026212.1 580 Missense Mutation ATG,GTG M157V XP_016881701.1
XM_017026213.1 580 Intron XP_016881702.1

View Full Product Details