Product Details

SNP ID
rs146328123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8302836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGACTGGTCTCCGGCAGAGGAG[G/T]ATGTGGCATTCCCGACAGAGGGGAC
Phenotype
MIM: 606475 MIM: 602139
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CD320 PubMed Links

Gene Details

Gene
CD320
Gene Name
CD320 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165895.1 713 Missense Mutation TAC,TCC Y174S NP_001159367.1
NM_016579.3 713 Missense Mutation TAC,TCC Y216S NP_057663.1
Gene
NDUFA7
Gene Name
NADH:ubiquinone oxidoreductase subunit A7
There are no transcripts associated with this gene.

View Full Product Details