Product Details

SNP ID
rs146423097
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40741867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTCCCAGATGGGCTTTAAAACT[A/G]TGAAGACATGAGGTGGGGGAGGGGT
Phenotype
MIM: 606476 MIM: 182285
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C19orf54 PubMed Links

Gene Details

Gene
C19orf54
Gene Name
chromosome 19 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198476.3 2198 UTR 3 NP_940878.3
XM_005258776.4 2198 UTR 3 XP_005258833.1
XM_005258778.4 2198 UTR 3 XP_005258835.1
XM_006723152.2 2198 Intron XP_006723215.1
XM_006723153.2 2198 Intron XP_006723216.1
XM_011526774.2 2198 Intron XP_011525076.1
XM_011526775.1 2198 Intron XP_011525077.1
XM_011526776.2 2198 Intron XP_011525078.1
XM_011526777.2 2198 Intron XP_011525079.1
XM_011526778.2 2198 Intron XP_011525080.1
XM_011526779.2 2198 Intron XP_011525081.1
XM_011526782.2 2198 Intron XP_011525084.1
XM_011526783.2 2198 Intron XP_011525085.1
XM_011526784.2 2198 Intron XP_011525086.1
XM_011526785.2 2198 Intron XP_011525087.1
XM_011526786.2 2198 Intron XP_011525088.1
XM_017026620.1 2198 Intron XP_016882109.1
XM_017026621.1 2198 Intron XP_016882110.1
XM_017026622.1 2198 Intron XP_016882111.1
XM_017026623.1 2198 Intron XP_016882112.1
XM_017026624.1 2198 Intron XP_016882113.1
XM_017026625.1 2198 Intron XP_016882114.1
XM_017026626.1 2198 UTR 3 XP_016882115.1
XM_017026627.1 2198 UTR 3 XP_016882116.1
Gene
ITPKC
Gene Name
inositol-trisphosphate 3-kinase C
There are no transcripts associated with this gene.

Gene
SNRPA
Gene Name
small nuclear ribonucleoprotein polypeptide A
There are no transcripts associated with this gene.

View Full Product Details